Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-methylphenyl isocyanate, 98%


Quantity 1g

Manufacturer: alfa aesar™  L1109503

 View more versions of this product

Catalog No. AAL1109503

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 212.05
MDL Number MFCD00037039
Molecular Formula C8H6BrNO
CAS 1591-98-6
Health Hazard 1 Danger
Chemical Name or Material 4-Bromo-2-methylphenyl isocyanate
Assay 98%
Melting Point 47° to 51°C
Beilstein 7633645
Odor Irritating
Health Hazard 3 GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point >110°C (230°F)
Health Hazard 2 GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
UN Number UN2811
Quantity 1g
Recommended Storage 2° to 8°C

Specifications

Chemical Name or Material 4-Bromo-2-methylphenyl isocyanate
CAS 1591-98-6
Molecular Formula C8H6BrNO
Formula Weight 212.05
MDL Number MFCD00037039
UN Number UN2811
Beilstein 7633645
Accessories

Danger

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C

SDS & Safety Documents