Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-methylphenyl isocyanate, 98%

Chemical Identifiers

Chemical Name or Material 4-Bromo-2-methylphenyl isocyanate
CAS 1591-98-6
Molecular Formula C8H6BrNO
Formula Weight 212.05
MDL Number MFCD00037039
UN Number UN2811
Beilstein 7633645
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1109506

 
alfa aesar™
L1109506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1109503

 
alfa aesar™
L1109503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 212.05
Chemical Name or Material 4-Bromo-2-methylphenyl isocyanate
Molecular Formula C8H6BrNO
CAS 1591-98-6
Assay 98%
MDL Number MFCD00037039
UN Number UN2811
Beilstein 7633645
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Melting Point 47° to 51°C
Quantity 5g
Flash Point >110°C (230°F)
Odor Irritating
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C