missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-methylphenyl isocyanate, 98%

Manufacturer:  Alfa Aesar™ L1109506

 View more versions of this product

Catalog No. AAL1109506



Specifications

Specifications

4-Bromo-2-methylphenyl isocyanate
Irritating
5g
C8H6BrNO
UN2811
4-bromo-2-methylphenyl isocyanate, benzene,4-bromo-1-isocyanato-2-methyl, 4-bromo-2-methylphenylisocyanate, 4-bromo-2-methylbenzenisocyanate, acmc-1bpqh, 5-bromo-2-isocyanatotoluene, 2-methyl-4-bromophenyl isocyanate, 4-bromanyl-1-isocyanato-2-methyl-benzene
CC1=C(C=CC(=C1)Br)N=C=O
212.046
212.05
>110°C (230°F)
47°C to 51°C
1591-98-6
MFCD00037039
7633645
NXAYRHQYINQHRF-UHFFFAOYSA-N
4-bromo-1-isocyanato-2-methylbenzene
2735594
98%

Chemical Identifiers

1591-98-6
212.046
NXAYRHQYINQHRF-UHFFFAOYSA-N
2735594
CC1=C(C=CC(=C1)Br)N=C=O
C8H6BrNO
MFCD00037039
4-bromo-2-methylphenyl isocyanate, benzene,4-bromo-1-isocyanato-2-methyl, 4-bromo-2-methylphenylisocyanate, 4-bromo-2-methylbenzenisocyanate, acmc-1bpqh, 5-bromo-2-isocyanatotoluene, 2-methyl-4-bromophenyl isocyanate, 4-bromanyl-1-isocyanato-2-methyl-benzene
4-bromo-1-isocyanato-2-methylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Recommended Storage : 2° to 8°C

Documents