missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-2-methylbenzonitrile, TCI America™

$45.83 - $158.89

Chemical Identifiers

CAS 67832-11-5
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.047
MDL Number MFCD00041461
InChI Key LPEBMDFRIKYFCF-UHFFFAOYSA-N
Synonym benzonitrile, 4-bromo-2-methyl, 5-bromo-2-cyanotoluene, 4-bromo-2-methyl-benzonitrile, 2-methyl-4-bromo benzonitrile, 2-methyl-4-bromobenzonitrile, 4-bromo-2-methylbenzene carbonitrile, 4-bromo-2-methylbenzenecarbonitrile, pubchem4721, 4-bromo-methylbenzonitrile, 4bromo-2-methylbenzonitrile
PubChem CID 144267
IUPAC Name 4-bromo-2-methylbenzonitrile
SMILES CC1=C(C=CC(=C1)Br)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B39935G View Documents TCI America
B39935G
5g Each for $45.83
 
B399325G View Documents TCI America
B399325G
25g Each for $158.89
 
Specifications

Chemical Identifiers

67832-11-5
196.047
LPEBMDFRIKYFCF-UHFFFAOYSA-N
144267
CC1=C(C=CC(=C1)Br)C#N
C8H6BrN
MFCD00041461
benzonitrile, 4-bromo-2-methyl, 5-bromo-2-cyanotoluene, 4-bromo-2-methyl-benzonitrile, 2-methyl-4-bromo benzonitrile, 2-methyl-4-bromobenzonitrile, 4-bromo-2-methylbenzene carbonitrile, 4-bromo-2-methylbenzenecarbonitrile, pubchem4721, 4-bromo-methylbenzonitrile, 4bromo-2-methylbenzonitrile
4-bromo-2-methylbenzonitrile

Specifications

White
5g
C8H6BrN
3439
LPEBMDFRIKYFCF-UHFFFAOYSA-N
4-bromo-2-methylbenzonitrile
144267
Crystal-Powder at 20°C
4-Bromo-2-methylbenzonitrile
67°C
67832-11-5
MFCD00041461
benzonitrile, 4-bromo-2-methyl, 5-bromo-2-cyanotoluene, 4-bromo-2-methyl-benzonitrile, 2-methyl-4-bromo benzonitrile, 2-methyl-4-bromobenzonitrile, 4-bromo-2-methylbenzene carbonitrile, 4-bromo-2-methylbenzenecarbonitrile, pubchem4721, 4-bromo-methylbenzonitrile, 4bromo-2-methylbenzonitrile
CC1=C(C=CC(=C1)Br)C#N
196.047
196.05
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No