missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-methylbenzonitrile, 98%

Manufacturer:  Alfa Aesar™ L1052603

 View more versions of this product

Catalog No. AAL1052603



Specifications

Specifications

4-Bromo-2-methylbenzonitrile
1g
C8H6BrN
UN3439
benzonitrile, 4-bromo-2-methyl, 5-bromo-2-cyanotoluene, 4-bromo-2-methyl-benzonitrile, 2-methyl-4-bromo benzonitrile, 2-methyl-4-bromobenzonitrile, 4-bromo-2-methylbenzene carbonitrile, 4-bromo-2-methylbenzenecarbonitrile, pubchem4721, 4-bromo-methylbenzonitrile, 4bromo-2-methylbenzonitrile
CC1=C(C=CC(=C1)Br)C#N
196.047
196.05
63°C to 64°C
67832-11-5
MFCD00041461
2574103
LPEBMDFRIKYFCF-UHFFFAOYSA-N
4-bromo-2-methylbenzonitrile
144267
98%

Chemical Identifiers

67832-11-5
196.047
LPEBMDFRIKYFCF-UHFFFAOYSA-N
144267
CC1=C(C=CC(=C1)Br)C#N
C8H6BrN
MFCD00041461
benzonitrile, 4-bromo-2-methyl, 5-bromo-2-cyanotoluene, 4-bromo-2-methyl-benzonitrile, 2-methyl-4-bromo benzonitrile, 2-methyl-4-bromobenzonitrile, 4-bromo-2-methylbenzene carbonitrile, 4-bromo-2-methylbenzenecarbonitrile, pubchem4721, 4-bromo-methylbenzonitrile, 4bromo-2-methylbenzonitrile
4-bromo-2-methylbenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Documents