missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Bromo-2-methylbenzene-1-sulfonyl chloride, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 451780100
Specifications
4-Bromo-2-methylbenzene-1-sulfonyl chloride | |
164.0°C (15.0 mmHg) | |
C7H6BrClO2S | |
10g | |
KNBSFUSBZNMAKU-UHFFFAOYSA-N | |
4-bromo-2-methylbenzenesulfonyl chloride | |
2781338 | |
97% |
139937-37-4 | |
96% min. (GC) | |
Br(CH3)C6H3SO2Cl | |
4-bromo-2-methylbenzene-1-sulfonyl chloride, 4-bromo-2-methylbenzenesulphonyl chloride, 4-bromo-2-methyl-benzenesulfonyl chloride, 2-methyl-4-bromobenzenesulfonylchloride, 5-bromo-2-chlorosulphonyl toluene, 2-methyl-4-bromobenzenesulfonyl chloride, benzenesulfonylchloride, 4-bromo-2-methyl, pubchem5066, buttpark 9857-56, intermediates-zcf02072 | |
CC1=C(C=CC(=C1)Br)S(=O)(=O)Cl | |
269.55 | |
269.55 |
Chemical Identifiers
139937-37-4 | |
269.55 | |
4-bromo-2-methylbenzene-1-sulfonyl chloride, 4-bromo-2-methylbenzenesulphonyl chloride, 4-bromo-2-methyl-benzenesulfonyl chloride, 2-methyl-4-bromobenzenesulfonylchloride, 5-bromo-2-chlorosulphonyl toluene, 2-methyl-4-bromobenzenesulfonyl chloride, benzenesulfonylchloride, 4-bromo-2-methyl, pubchem5066, buttpark 9857-56, intermediates-zcf02072 | |
4-bromo-2-methylbenzenesulfonyl chloride |
C7H6BrClO2S | |
KNBSFUSBZNMAKU-UHFFFAOYSA-N | |
2781338 | |
CC1=C(C=CC(=C1)Br)S(=O)(=O)Cl |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
RUO â Research Use Only