missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-methylbenzaldehyde, 95%

Manufacturer:  Alfa Aesar™ H61314MD

 View more versions of this product

Catalog No. AAH61314MD



Specifications

Specifications

4-Bromo-2-methylbenzaldehyde
24078-12-4
MFCD07787171
RCBPVESMGNZMSG-UHFFFAOYSA-N
4-bromo-2-methylbenzaldehyde
13555150
Solid
250mg
C8H7BrO
2-methyl-4-bromobenzaldehyde
CC1=C(C=CC(=C1)Br)C=O
199.047
199.04
95%

Chemical Identifiers

24078-12-4
199.047
RCBPVESMGNZMSG-UHFFFAOYSA-N
13555150
CC1=C(C=CC(=C1)Br)C=O
C8H7BrO
MFCD07787171
2-methyl-4-bromobenzaldehyde
4-bromo-2-methylbenzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents