missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Bromo-2-methylaniline, 97%, ACROS Organics™
Chemical Identifiers
CAS | 583-75-5 |
---|---|
Molecular Formula | C7H8BrN |
Molecular Weight (g/mol) | 186.05 |
MDL Number | MFCD00007825 |
InChI Key | PCHYYOCUCGCSBU-UHFFFAOYSA-N |
Synonym | 1-amino-4-bromo-2-methylbenzene, 2-amino-5-bromotoluene, 2-methyl-4-bromoaniline, 4-bromo-2-methyl-phenylamine, 4-bromo-2-methylbenenamine, 4-bromo-2-methylbenzenamine, 4-bromo-o-toluidine, 5-bromo-2-aminotoluene, benzenamine, 4-bromo-2-methyl, o-toluidine, 4-bromo |
PubChem CID | 11423 |
IUPAC Name | 4-bromo-2-methylaniline |
SMILES | CC1=C(C=CC(=C1)Br)N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC167490250
|
Thermo Fisher Scientific
167490250 |
25g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
583-75-5 | |
186.05 | |
PCHYYOCUCGCSBU-UHFFFAOYSA-N | |
11423 | |
CC1=C(C=CC(=C1)Br)N |
C7H8BrN | |
MFCD00007825 | |
1-amino-4-bromo-2-methylbenzene, 2-amino-5-bromotoluene, 2-methyl-4-bromoaniline, 4-bromo-2-methyl-phenylamine, 4-bromo-2-methylbenenamine, 4-bromo-2-methylbenzenamine, 4-bromo-o-toluidine, 5-bromo-2-aminotoluene, benzenamine, 4-bromo-2-methyl, o-toluidine, 4-bromo | |
4-bromo-2-methylaniline |
Specifications
>112°C | |
Glass bottle | |
Beige to Brown-Purple | |
25g | |
96.0 | |
96% min. (GC) | |
BrC6H3(CH3)NH2 | |
12, 838 | |
PCHYYOCUCGCSBU-UHFFFAOYSA-N | |
4-bromo-2-methylaniline | |
11423 | |
Crystalline Powder | |
4-Bromo-2-methylaniline, 97% |
Authentic | |
240°C | |
53°C to 58°C | |
583-75-5 | |
100.0 | |
C7H8BrN | |
MFCD00007825 | |
1-amino-4-bromo-2-methylbenzene, 2-amino-5-bromotoluene, 2-methyl-4-bromoaniline, 4-bromo-2-methyl-phenylamine, 4-bromo-2-methylbenenamine, 4-bromo-2-methylbenzenamine, 4-bromo-o-toluidine, 5-bromo-2-aminotoluene, benzenamine, 4-bromo-2-methyl, o-toluidine, 4-bromo | |
CC1=C(C=CC(=C1)Br)N | |
186.05 | |
186.05 | |
97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protecti
GHS Signal Word: Warning
EINECSNumber : 209-519-9
RTECSNumber : XU4250000
TSCA : TSCA