Please login to your online account to display your discounted pricing

4-Bromo-2-methylaniline, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  167490250

 View more versions of this product

Catalog No. AC167490250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 186.05
Physical Form Crystalline Powder
Quantity 25g
Color Beige to purple-brown
Beilstein 12, 838
TSCA TSCA
RTECS Number XU4250000
EINECS Number 209-519-9
Melting Point 53° to 58°C
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
Linear Formula BrC6H3(CH3)NH2
Assay Percent Range 97%
CAS 583-75-5
Chemical Name or Material 4-Bromo-2-methylaniline
Molecular Formula C7H8BrN
MDL Number MFCD00007825
Name Note 97%
Packaging Glass bottle
Synonym 4-Bromo-o-toluidine
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Bromo-2-methylaniline
Name Note 97%
CAS 583-75-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H8BrN
Linear Formula BrC6H3(CH3)NH2
Formula Weight 186.05
MDL Number MFCD00007825
Synonym 4-Bromo-o-toluidine
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.

SDS & Safety Documents