missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4'-Bromo-2'-methylacetanilide, 99+%, ACROS Organics™

Chemical Identifiers

CAS 24106-05-6
Molecular Formula C9H10BrNO
Molecular Weight (g/mol) 228.089
MDL Number MFCD00017779
InChI Key IJZSFOIZVXCSFP-UHFFFAOYSA-N
Synonym 2-acetamido-5-bromotoluene, 2-methyl-4-bromoacetanilide, 4'-bromo-2'-methylacetanilide, 4'-bromo-o-acetotoluidide, 4'-bromoaceto-o-toluidide, 4-bromo-2-methyl acetanilide, 4-bromo-2-methylacetanilide, acmc-1cjyt, n-4-bromo-2-methylphenyl acetamide, o-acetotoluidide, 4'-bromo
PubChem CID 222277
IUPAC Name N-(4-bromo-2-methylphenyl)acetamide
SMILES CC1=C(C=CC(=C1)Br)NC(=O)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC263380250
SDS View Documents
Thermo Fisher Scientific
263380250
25g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

24106-05-6
228.089
IJZSFOIZVXCSFP-UHFFFAOYSA-N
222277
CC1=C(C=CC(=C1)Br)NC(=O)C
C9H10BrNO
MFCD00017779
2-acetamido-5-bromotoluene, 2-methyl-4-bromoacetanilide, 4'-bromo-2'-methylacetanilide, 4'-bromo-o-acetotoluidide, 4'-bromoaceto-o-toluidide, 4-bromo-2-methyl acetanilide, 4-bromo-2-methylacetanilide, acmc-1cjyt, n-4-bromo-2-methylphenyl acetamide, o-acetotoluidide, 4'-bromo
N-(4-bromo-2-methylphenyl)acetamide

Specifications

Authentic
Beige-Brown to Pink
25g
99.0
99+%
MFCD00017779
IJZSFOIZVXCSFP-UHFFFAOYSA-N
N-(4-bromo-2-methylphenyl)acetamide
222277
Crystalline Powder
4';-Bromo-2';-methylacetanilide, 99+%
Glass Bottle
159°C to 162°C
24106-05-6
100.0
C9H10BrNO
2-acetamido-5-bromotoluene, 2-methyl-4-bromoacetanilide, 4'-bromo-2'-methylacetanilide, 4'-bromo-o-acetotoluidide, 4'-bromoaceto-o-toluidide, 4-bromo-2-methyl acetanilide, 4-bromo-2-methylacetanilide, acmc-1cjyt, n-4-bromo-2-methylphenyl acetamide, o-acetotoluidide, 4'-bromo
CC1=C(C=CC(=C1)Br)NC(=O)C
228.089
228.1
≥99.0%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

TSCA : TSCA

SDS