missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4'-Bromo-2'-methylacetanilide, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 263380050

 View more versions of this product

Catalog No. AC263380050


 

Specifications Description & Specifications

Specifications

4';-Bromo-2';-methylacetanilide
24106-05-6
100.0
Glass bottle
5g
Crystalline Powder
99+%
228.1
99+%
99.0
Authentic
159° to 162°C
Pale pink to light beige-brown
≥99.0%
C9H10BrNO
MFCD00017779

Chemical Identifiers

24106-05-6
100.0
C9H10BrNO
MFCD00017779
≥99.0%
99.0
99+%
228.1
4';-Bromo-2';-methylacetanilide, 99+%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

TSCA : TSCA