Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-methoxybenzyl alcohol, 97%

$32.18 - $335.57

Chemical Identifiers

Chemical Name or Material 4-Bromo-2-methoxybenzyl alcohol
CAS 17102-63-5
Molecular Formula C8H9BrO2
Formula Weight 217.06
MDL Number MFCD07369762
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2733903

 
alfa aesar™
H2733903
1g Each for $32.18

AAH2733906

 
alfa aesar™
H2733906
5g Each for $107.43

AAH2733914

 
alfa aesar™
H2733914
25g Each for $335.57
Description & Specifications

Specifications

Formula Weight 217.06
Chemical Name or Material 4-Bromo-2-methoxybenzyl alcohol
Assay 97%
Molecular Formula C8H9BrO2
MDL Number MFCD07369762
CAS 17102-63-5
Melting Point 66° to 70°C
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
It is used in azaquinone-methide-mediated depolymerization of aromatic carbamate oligomers.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.