missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-2-iodoaniline, TCI America™

$76.39 - $230.70

Chemical Identifiers

CAS 66416-72-6
Molecular Formula C6H5BrIN
Molecular Weight (g/mol) 297.921
MDL Number MFCD02093764
InChI Key HHTYEQWCHQEJNV-UHFFFAOYSA-N
Synonym 4-bromo-2-iodo-phenylamine, 4-bromo-2-iodobenzenamine, 2-iodo-4-bromoaniline, benzenamine,4-bromo-2-iodo, benzenamine, 4-bromo-2-iodo, pubchem3784, acmc-1b9hb, 4-bromo-2-iodophenylamine, asischem t54343, 4-bromo-2-iodo-benzenamine
PubChem CID 823934
IUPAC Name 4-bromo-2-iodoaniline
SMILES C1=CC(=C(C=C1Br)I)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B31641G View Documents TCI America
B31641G
1g Each for $76.39
 
B31645G View Documents TCI America
B31645G
5g Each for $230.70
 
Specifications

Chemical Identifiers

66416-72-6
297.921
HHTYEQWCHQEJNV-UHFFFAOYSA-N
823934
C1=CC(=C(C=C1Br)I)N
C6H5BrIN
MFCD02093764
4-bromo-2-iodo-phenylamine, 4-bromo-2-iodobenzenamine, 2-iodo-4-bromoaniline, benzenamine,4-bromo-2-iodo, benzenamine, 4-bromo-2-iodo, pubchem3784, acmc-1b9hb, 4-bromo-2-iodophenylamine, asischem t54343, 4-bromo-2-iodo-benzenamine
4-bromo-2-iodoaniline

Specifications

Purple-White
1g
C6H5BrIN
2811
HHTYEQWCHQEJNV-UHFFFAOYSA-N
4-bromo-2-iodoaniline
823934
Crystal-Powder at 20°C
4-Bromo-2-iodoaniline
70°C
66416-72-6
MFCD02093764
4-bromo-2-iodo-phenylamine, 4-bromo-2-iodobenzenamine, 2-iodo-4-bromoaniline, benzenamine,4-bromo-2-iodo, benzenamine, 4-bromo-2-iodo, pubchem3784, acmc-1b9hb, 4-bromo-2-iodophenylamine, asischem t54343, 4-bromo-2-iodo-benzenamine
C1=CC(=C(C=C1Br)I)N
297.921
297.92
≥97.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : No