missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-2-hydroxypyridine, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC432090010

 View more versions of this product

Catalog No. AC432090010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

4-Bromo-2-hydroxypyridine
96.0
Authentic
White
≥96.0%
1g
174
36953-37-4
100.0
Glass bottle
Crystalline Powder
97%
C5H4BrNO
MFCD00234041

Chemical Identifiers

36953-37-4
100.0
C5H4BrNO
MFCD00234041
≥96.0%
96.0
97%
174
4-Bromo-2-hydroxypyridine

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.