missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-2-hydroxypyridine, 97%, ACROS Organics™

Chemical Identifiers

CAS 36953-37-4
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C5H4BrNO
Formula Weight 174
MDL Number MFCD00234041
Chemical Name or Material 4-Bromo-2-hydroxypyridine
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC432090050 SDS Thermo Fisher Scientific
AC432090050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC432090010 SDS Thermo Fisher Scientific
AC432090010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

36953-37-4
100.0
C5H4BrNO
MFCD00234041
≥96.0%
96.0
97%
174
4-Bromo-2-hydroxypyridine

Specifications

Crystalline Powder
5g
96.0
36953-37-4
MFCD00234041
97%
4-Bromo-2-hydroxypyridine
174
White
100.0
Authentic
C5H4BrNO
Glass bottle
≥96.0%

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.