missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4'-Bromo-2'-hydroxy-5'-methylacetophenone, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2912506

 View more versions of this product

Catalog No. AAH2912506



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4'-Bromo-2'-hydroxy-5'-methylacetophenone
86°C to 89°C
MFCD08273792
1-4-bromo-2-hydroxy-5-methylphenyl ethanone, 2-acetyl-4-methyl-5-bromophenol, 4'-bromo-2'-hydroxy-5'-methylacetophenone, 1-4-bromo-2-hydroxy-5-methylphenyl ethan-1-one, 1-acetyl-4-bromo-2-hydroxy-5-methylbenzene, 4'-bromo-2'-hydroxy-5'-methyl acetophenone
CC1=C(C=C(C(=C1)C(=O)C)O)Br
229.073
229.07
1020253-81-9
C9H9BrO2
5 g
BPQABGQSVOUODJ-UHFFFAOYSA-N
1-(4-bromo-2-hydroxy-5-methylphenyl)ethanone
42553093
97%

Chemical Identifiers

1020253-81-9
229.073
BPQABGQSVOUODJ-UHFFFAOYSA-N
42553093
CC1=C(C=C(C(=C1)C(=O)C)O)Br
C9H9BrO2
MFCD08273792
1-4-bromo-2-hydroxy-5-methylphenyl ethanone, 2-acetyl-4-methyl-5-bromophenol, 4'-bromo-2'-hydroxy-5'-methylacetophenone, 1-4-bromo-2-hydroxy-5-methylphenyl ethan-1-one, 1-acetyl-4-bromo-2-hydroxy-5-methylbenzene, 4'-bromo-2'-hydroxy-5'-methyl acetophenone
1-(4-bromo-2-hydroxy-5-methylphenyl)ethanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only