Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-furaldehyde, 97%

$54.55 - $138.93

Chemical Identifiers

Chemical Name or Material 4-Bromo-2-furaldehyde
CAS 21921-76-6
Molecular Formula C5H3BrO2
Formula Weight 174.98
MDL Number MFCD00671504
Synonym 4-Bromofuran-2-carboxaldehyde; 4-Bromofurfural
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2699403

  SDS
alfa aesar™
H2699403
1g Each for $138.93

AAH26994MD

  SDS
alfa aesar™
H26994MD
250mg Each for $54.55
Description & Specifications

Specifications

Formula Weight 174.98
Chemical Name or Material 4-Bromo-2-furaldehyde
Melting Point 54° to 58°C
Assay 97%
Quantity 1g
Molecular Formula C5H3BrO2
MDL Number MFCD00671504
CAS 21921-76-6
Synonym 4-Bromofuran-2-carboxaldehyde; 4-Bromofurfural
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C