missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-2-fluoropyridine, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC442820050

 View more versions of this product

Catalog No. AC442820050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Bromo-2-fluoropyridine
Authentic
65.0°C (5.0 Torr)
5g
96.0
96% min. (GC)
2-fluoro-4-bromopyridine, 4-bromanyl-2-fluoranyl-pyridine, 4-bromo-2-fluoro pyridine, 4-bromo-2-fluoro-pyridine, abbypharma ap-17-5099, acmc-209bdf, ksc493i4r, pubchem1228, pyff21, pyridine, 4-bromo-2-fluoro
C1=CN=C(C=C1Br)F
175.99
175.99
97%
71°C
Glass bottle
Colorless to Yellow
128071-98-7
100.0
C5H3BrFN
PTPTZLXZHPPVKG-UHFFFAOYSA-N
4-bromo-2-fluoropyridine
2762851
Liquid

Chemical Identifiers

128071-98-7
175.99
2-fluoro-4-bromopyridine, 4-bromanyl-2-fluoranyl-pyridine, 4-bromo-2-fluoro pyridine, 4-bromo-2-fluoro-pyridine, abbypharma ap-17-5099, acmc-209bdf, ksc493i4r, pubchem1228, pyff21, pyridine, 4-bromo-2-fluoro
4-bromo-2-fluoropyridine
C5H3BrFN
PTPTZLXZHPPVKG-UHFFFAOYSA-N
2762851
C1=CN=C(C=C1Br)F
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

SDS Documents