Learn More
4-Bromo-2-fluorophenol, 98%, Thermo Scientific Chemicals
$147.85 - $147.85
Chemical Identifiers
CAS | 2105-94-4 |
---|---|
Molecular Formula | C6H4BrFO |
Molecular Weight (g/mol) | 191.00 |
MDL Number | MFCD00011722 |
InChI Key | RYVOZMPTISNBDB-UHFFFAOYSA-N |
Synonym | 2-fluoro-4-bromophenol, 4-bromo-2-fluoro-phenol, phenol, 4-bromo-2-fluoro, 4-bromo-2-fluoro phenol, pubchem1488, p-bromo-o-fluorophenol, 2-fluoro-4-bromo phenol, acmc-209fh4, ksc204a3n, 3-fluoro-4-hydroxy-bromobenzene |
PubChem CID | 2724981 |
IUPAC Name | 4-bromo-2-fluorophenol |
SMILES | OC1=CC=C(Br)C=C1F |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC293920250
|
Thermo Scientific Chemicals
293920250 |
25 g | Glass bottle |
Each for $147.85
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2105-94-4 | |
191.00 | |
RYVOZMPTISNBDB-UHFFFAOYSA-N | |
2724981 | |
OC1=CC=C(Br)C=C1F |
C6H4BrFO | |
MFCD00011722 | |
2-fluoro-4-bromophenol, 4-bromo-2-fluoro-phenol, phenol, 4-bromo-2-fluoro, 4-bromo-2-fluoro phenol, pubchem1488, p-bromo-o-fluorophenol, 2-fluoro-4-bromo phenol, acmc-209fh4, ksc204a3n, 3-fluoro-4-hydroxy-bromobenzene | |
4-bromo-2-fluorophenol |
Specifications
2105-94-4 | |
100.0 | |
1.7400g/mL | |
98°C | |
97.5% min. (GC) | |
C6H4BrFO | |
BrC6H3(F)OH | |
25 g | |
2-fluoro-4-bromophenol, 4-bromo-2-fluoro-phenol, phenol, 4-bromo-2-fluoro, 4-bromo-2-fluoro phenol, pubchem1488, p-bromo-o-fluorophenol, 2-fluoro-4-bromo phenol, acmc-209fh4, ksc204a3n, 3-fluoro-4-hydroxy-bromobenzene | |
OC1=CC=C(Br)C=C1F | |
191.00 | |
191 | |
Liquid |
97.5 | |
Colorless to Brown | |
79°C (7.0 mmHg) | |
Authentic | |
Glass bottle | |
1.5660 to 1.568 | |
MFCD00011722 | |
1.74 | |
RYVOZMPTISNBDB-UHFFFAOYSA-N | |
4-bromo-2-fluorophenol | |
2724981 | |
98% | |
4-Bromo-2-fluorophenol |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only