missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-2-fluorophenol, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 293920050

 View more versions of this product

Catalog No. AC293920050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

4-Bromo-2-fluorophenol
98°C
Glass bottle
1.74
Colorless to Brown
2105-94-4
100.0
C6H4BrFO
MFCD00011722
RYVOZMPTISNBDB-UHFFFAOYSA-N
4-bromo-2-fluorophenol
2724981
Liquid
1.7400g/mL
Authentic
1.5660 to 1.568
79°C (7.0mmHg)
5g
97.5
97.5% min. (GC)
BrC6H3(F)OH
2-fluoro-4-bromo phenol, 2-fluoro-4-bromophenol, 3-fluoro-4-hydroxy-bromobenzene, 4-bromo-2-fluoro phenol, 4-bromo-2-fluoro-phenol, acmc-209fh4, ksc204a3n, p-bromo-o-fluorophenol, phenol, 4-bromo-2-fluoro, pubchem1488
C1=CC(=C(C=C1Br)F)O
191
191
98%

Chemical Identifiers

2105-94-4
191
RYVOZMPTISNBDB-UHFFFAOYSA-N
2724981
C1=CC(=C(C=C1Br)F)O
C6H4BrFO
MFCD00011722
2-fluoro-4-bromo phenol, 2-fluoro-4-bromophenol, 3-fluoro-4-hydroxy-bromobenzene, 4-bromo-2-fluoro phenol, 4-bromo-2-fluoro-phenol, acmc-209fh4, ksc204a3n, p-bromo-o-fluorophenol, phenol, 4-bromo-2-fluoro, pubchem1488
4-bromo-2-fluorophenol
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents