Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-fluorocinnamic acid, 98%

$55.42 - $230.89

Chemical Identifiers

Chemical Name or Material 4-Bromo-2-fluorocinnamic acid
CAS 149947-19-3
Molecular Formula C9H6BrFO2
Formula Weight 245.05
MDL Number MFCD00143267
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1602806

  SDS
alfa aesar™
A1602806
5g Each for $55.42

AAA1602814

  SDS
alfa aesar™
A1602814
25g Each for $230.89
Description & Specifications

Specifications

Formula Weight 245.05
Chemical Name or Material 4-Bromo-2-fluorocinnamic acid
Assay 98%
Molecular Formula C9H6BrFO2
MDL Number MFCD00143267
CAS 149947-19-3
Melting Point 216° to 221°C
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.