Please login to your online account to display your discounted pricing

4-Bromo-2-fluorocinnamic acid, 97%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  AC290550050

 View more versions of this product

Catalog No. AC290550050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Fine Crystalline Powder
Assay ≥96.0%
Formula Weight 245.05
Quantity 5g
Color Light green
Melting Point 219° to 223°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 4-Bromo-2-fluorocinnamic acid
Name Note 97%
Assay Percent Range 97%
Molecular Formula C9H6BrFO2
MDL Number MFCD00143267
CAS 149947-19-3
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 4-Bromo-2-fluorocinnamic acid
Name Note 97%
CAS 149947-19-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H6BrFO2
Formula Weight 245.05
MDL Number MFCD00143267
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents