Please login to your online account to display your discounted pricing

4-Bromo-2-fluorocinnamic acid, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Bromo-2-fluorocinnamic acid
Name Note 97%
CAS 149947-19-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H6BrFO2
Formula Weight 245.05
MDL Number MFCD00143267
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC290550050

  SDS
thermo fisher scientific
AC290550050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Fine Crystalline Powder
Assay ≥96.0%
Formula Weight 245.05
Quantity 5g
Color Light green
Melting Point 219° to 223°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 4-Bromo-2-fluorocinnamic acid
Name Note 97%
Assay Percent Range 97%
Molecular Formula C9H6BrFO2
MDL Number MFCD00143267
CAS 149947-19-3
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.