missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-2-fluorobiphenyl, TCI America™

$42.78 - $548.48

Chemical Identifiers

CAS 41604-19-7
Molecular Formula C12H8BrF
Molecular Weight (g/mol) 251.098
MDL Number MFCD00051716
InChI Key HTRNHWBOBYFTQF-UHFFFAOYSA-N
Synonym 4-bromo-2-fluorobiphenyl, 4-bromo-2-fluoro-1,1'-biphenyl, 2-fluoro-4-bromo biphenyl, 1,1'-biphenyl, 4-bromo-2-fluoro, 4-bromo-2-fluorodiphenyl, 4-bromo-2-fluoro-1,1-biphenyl, 4-bromo-2-fluoro biphenyl, pubchem2652, acmc-209jkv, 2-fluoro-4-bromobiphenyl
PubChem CID 521063
IUPAC Name 4-bromo-2-fluoro-1-phenylbenzene
SMILES C1=CC=C(C=C1)C2=C(C=C(C=C2)Br)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B20285G View Documents TCI America
B20285G
5g Each for $42.78
 
B2028250G View Documents TCI America
B2028250G
250g Each for $548.48
 
B202825G View Documents TCI America
B202825G
25g Each for $103.89
 
Specifications

Chemical Identifiers

41604-19-7
251.098
HTRNHWBOBYFTQF-UHFFFAOYSA-N
521063
C1=CC=C(C=C1)C2=C(C=C(C=C2)Br)F
C12H8BrF
MFCD00051716
4-bromo-2-fluorobiphenyl, 4-bromo-2-fluoro-1,1'-biphenyl, 2-fluoro-4-bromo biphenyl, 1,1'-biphenyl, 4-bromo-2-fluoro, 4-bromo-2-fluorodiphenyl, 4-bromo-2-fluoro-1,1-biphenyl, 4-bromo-2-fluoro biphenyl, pubchem2652, acmc-209jkv, 2-fluoro-4-bromobiphenyl
4-bromo-2-fluoro-1-phenylbenzene

Specifications

White-Yellow
41604-19-7
MFCD00051716
4-bromo-2-fluorobiphenyl, 4-bromo-2-fluoro-1,1'-biphenyl, 2-fluoro-4-bromo biphenyl, 1,1'-biphenyl, 4-bromo-2-fluoro, 4-bromo-2-fluorodiphenyl, 4-bromo-2-fluoro-1,1-biphenyl, 4-bromo-2-fluoro biphenyl, pubchem2652, acmc-209jkv, 2-fluoro-4-bromobiphenyl
C1=CC=C(C=C1)C2=C(C=C(C=C2)Br)F
251.098
251.10
≥99.0% (GC)
5g
C12H8BrF
3152
HTRNHWBOBYFTQF-UHFFFAOYSA-N
4-bromo-2-fluoro-1-phenylbenzene
521063
Crystal-Lump at 20°C
4-Bromo-2-fluorobiphenyl
Safety and Handling

Safety and Handling

TSCA : No