missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-2-fluorobiphenyl, 98%, ACROS Organics™

Chemical Identifiers

CAS 41604-19-7
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C12H8BrF
Linear Formula C6H5C6H3(F)Br
Formula Weight 251.1
MDL Number MFCD00051716
Chemical Name or Material 4-Bromo-2-fluorobiphenyl
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC373270500 SDS Thermo Fisher Scientific
AC373270500
Glass bottle 50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC373270100 SDS Thermo Fisher Scientific
AC373270100
Glass Bottle 10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

41604-19-7
100.0
C12H8BrF
251.1
4-Bromo-2-fluorobiphenyl
97.5
98%
C6H5C6H3(F)Br
MFCD00051716
≥97.5%

Specifications

41604-19-7
100.0
Glass bottle
38° to 42°C
98%
C12H8BrF
251.1
4-Bromo-2-fluorobiphenyl
97.5
Authentic
Off-white to pale yellow
Crystalline Powder or Chunks
50g
C6H5C6H3(F)Br
MFCD00051716
≥97.5%

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning