missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-fluorobiphenyl, 98%

Manufacturer:  Alfa Aesar™ A1492414

 View more versions of this product

Catalog No. AAA1492414



Specifications

Specifications

4-Bromo-2-fluorobiphenyl
175°C (2mmHg)
25g
C12H8BrF
UN3152
4-bromo-2-fluorobiphenyl, 4-bromo-2-fluoro-1,1'-biphenyl, 2-fluoro-4-bromo biphenyl, 1,1'-biphenyl, 4-bromo-2-fluoro, 4-bromo-2-fluorodiphenyl, 4-bromo-2-fluoro-1,1-biphenyl, 4-bromo-2-fluoro biphenyl, pubchem2652, acmc-209jkv, 2-fluoro-4-bromobiphenyl
C1=CC=C(C=C1)C2=C(C=C(C=C2)Br)F
251.098
251.1
>110°C (230°F)
38°C to 41°C
41604-19-7
MFCD00051716
2208913
HTRNHWBOBYFTQF-UHFFFAOYSA-N
4-bromo-2-fluoro-1-phenylbenzene
521063
98%

Chemical Identifiers

41604-19-7
251.098
HTRNHWBOBYFTQF-UHFFFAOYSA-N
521063
C1=CC=C(C=C1)C2=C(C=C(C=C2)Br)F
C12H8BrF
MFCD00051716
4-bromo-2-fluorobiphenyl, 4-bromo-2-fluoro-1,1'-biphenyl, 2-fluoro-4-bromo biphenyl, 1,1'-biphenyl, 4-bromo-2-fluoro, 4-bromo-2-fluorodiphenyl, 4-bromo-2-fluoro-1,1-biphenyl, 4-bromo-2-fluoro biphenyl, pubchem2652, acmc-209jkv, 2-fluoro-4-bromobiphenyl
4-bromo-2-fluoro-1-phenylbenzene
Safety and Handling

Safety and Handling

DOTInformation : Transport Hazard Class: 9; Packing Group: II; Proper Shipping Name: POLYHALOGENATED BIPHENYLS, SOLID

SDS Documents