Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-fluorobiphenyl, 98%

Chemical Identifiers

Chemical Name or Material 4-Bromo-2-fluorobiphenyl
CAS 41604-19-7
Molecular Formula C12H8BrF
Formula Weight 251.1
MDL Number MFCD00051716
UN Number UN3152
Beilstein 2208913
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1492406

  SDS
alfa aesar™
A1492406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1492414

  SDS
alfa aesar™
A1492414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1492418

  SDS
alfa aesar™
A1492418
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 251.1
Assay 98%
Flash Point >110°C (230°F)
Beilstein 2208913
Quantity 5g
MDL Number MFCD00051716
Chemical Name or Material 4-Bromo-2-fluorobiphenyl
DOT Information Transport Hazard Class: 9; Packing Group: II; Proper Shipping Name: POLYHALOGENATED BIPHENYLS, SOLID
Boiling Point 175°C (2mmHg)
Molecular Formula C12H8BrF
CAS 41604-19-7
Melting Point 38° to 41°C
UN Number UN3152