Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-fluorobenzylamine hydrochloride, 98%

$82.97 - $917.41

Chemical Identifiers

Chemical Name or Material 4-Bromo-2-fluorobenzylamine hydrochloride
CAS 147181-08-6
Molecular Formula C7H7BrFN·HCl
Formula Weight 240.5
MDL Number MFCD00143266
Beilstein 6938847
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1565406

 
alfa aesar™
A1565406
5g Each for $82.97

AAA1565414

 
alfa aesar™
A1565414
25g Each for $281.69

AAA1565422

 
alfa aesar™
A1565422
100g Each for $917.41
Description & Specifications

Specifications

Formula Weight 240.5
Chemical Name or Material 4-Bromo-2-fluorobenzylamine hydrochloride
Assay 98%
Molecular Formula C7H7BrFN·HCl
MDL Number MFCD00143266
CAS 147181-08-6
Melting Point 254° to 257°C
Beilstein 6938847
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.