missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-fluorobenzylamine hydrochloride, 98%

Manufacturer:  Alfa Aesar™ A1565422

 View more versions of this product

Catalog No. AAA1565422



Specifications

Specifications

4-Bromo-2-fluorobenzylamine hydrochloride
100g
C7H8BrClFN
6938847
RTQOFJWBJNVJEO-UHFFFAOYSA-N
(4-bromo-2-fluorophenyl)methanamine;hydrochloride
18468197
98%
254°C to 257°C
147181-08-6
MFCD00143266
4-bromo-2-fluorobenzylamine hydrochloride, 4-bromo-2-fluorophenyl methanamine hydrochloride, 4-bromo-2-fluorobenzylamine hcl, 1-4-bromo-2-fluorophenyl methanamine hydrochloride, benzenemethanamine, 4-bromo-2-fluoro-, hydrochloride, 4-bromo-2-fluorobenzylamine hydrochlorid, pubchem3774, pubchem12660, acmc-1by0k, 4-bromo-2-fluoro-benzylamine hydrochloride
C1=CC(=C(C=C1Br)F)CN.Cl
240.5
240.5

Chemical Identifiers

147181-08-6
240.5
RTQOFJWBJNVJEO-UHFFFAOYSA-N
18468197
C1=CC(=C(C=C1Br)F)CN.Cl
C7H8BrClFN
MFCD00143266
4-bromo-2-fluorobenzylamine hydrochloride, 4-bromo-2-fluorophenyl methanamine hydrochloride, 4-bromo-2-fluorobenzylamine hcl, 1-4-bromo-2-fluorophenyl methanamine hydrochloride, benzenemethanamine, 4-bromo-2-fluoro-, hydrochloride, 4-bromo-2-fluorobenzylamine hydrochlorid, pubchem3774, pubchem12660, acmc-1by0k, 4-bromo-2-fluoro-benzylamine hydrochloride
(4-bromo-2-fluorophenyl)methanamine;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents