missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-bromo-2-fluorobenzylamine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 384440050

 View more versions of this product

Catalog No. AC384440050



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Bromo-2-fluorobenzylamine
97.5
Colorless to Yellow
97.5% min. (GC)
C7H7BrFN
NH2CH2C6H3(F)Br
5 g
RLTFBWCBGIZCDQ-UHFFFAOYSA-N
(4-bromo-2-fluorophenyl)methanamine
3770848
98%
112734-22-2
100.0
Authentic
Glass bottle
1.5630 to 1.5650 (20°C, 589nm)
MFCD00153076
4-bromo-2-fluorobenzylamine, 4-bromo-2-fluorophenyl methanamine, 2-fluoro-4-bromobenzylamine, 2-fluoro-4-bromobenzyl amine, 4-bromo-2-fluorobenzyl amine, benzenemethanamine, 4-bromo-2-fluoro, 1-4-bromo-2-fluorophenyl methanamine, 4-bromo-2-fluoro-phenyl methanamine, 4-bromo-2-fluorophenyl methylamine, pubchem4709
C1=CC(=C(C=C1Br)F)CN
204.04
204.04
Liquid

Chemical Identifiers

112734-22-2
204.04
RLTFBWCBGIZCDQ-UHFFFAOYSA-N
3770848
C1=CC(=C(C=C1Br)F)CN
C7H7BrFN
MFCD00153076
4-bromo-2-fluorobenzylamine, 4-bromo-2-fluorophenyl methanamine, 2-fluoro-4-bromobenzylamine, 2-fluoro-4-bromobenzyl amine, 4-bromo-2-fluorobenzyl amine, benzenemethanamine, 4-bromo-2-fluoro, 1-4-bromo-2-fluorophenyl methanamine, 4-bromo-2-fluoro-phenyl methanamine, 4-bromo-2-fluorophenyl methylamine, pubchem4709
(4-bromo-2-fluorophenyl)methanamine
Safety and Handling

Safety and Handling

GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face,hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only