missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-fluorobenzyl chloride, 97%

$52.58 - $171.18

Chemical Identifiers

CAS 85510-82-3
Molecular Formula C7H5BrClF
Molecular Weight (g/mol) 223.469
MDL Number MFCD02094036
InChI Key UDKQGFMDBMYVHI-UHFFFAOYSA-N
Synonym 4-bromo-2-fluorobenzyl chloride, 4-bromo-1-chloromethyl-2-fluorobenzene, 4-bromo-2-fluorobenzylchloride, 4-bromo-1-chloromethyl-2-fluoro-benzene, acmc-209q6a, 4-bromo-2-fluorobenzylchloride, 2-fluoro-4-bromo-chloromethylbenzene, 4-bromo-2-fluorobenzyl chloride, 4-bromo-2-fluorobenzyl chloride, 4-bromo-2-fluorobenzyl chloride
PubChem CID 2773361
IUPAC Name 4-bromo-1-(chloromethyl)-2-fluorobenzene
SMILES C1=CC(=C(C=C1Br)F)CCl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1868003 View Documents Alfa Aesar™
L1868003
1g Each for $52.58
 
AAL1868006 View Documents Alfa Aesar™
L1868006
5g Each for $171.18
 
Specifications

Chemical Identifiers

85510-82-3
223.469
UDKQGFMDBMYVHI-UHFFFAOYSA-N
2773361
C1=CC(=C(C=C1Br)F)CCl
C7H5BrClF
MFCD02094036
4-bromo-2-fluorobenzyl chloride, 4-bromo-1-chloromethyl-2-fluorobenzene, 4-bromo-2-fluorobenzylchloride, 4-bromo-1-chloromethyl-2-fluoro-benzene, acmc-209q6a, 4-bromo-2-fluorobenzylchloride, 2-fluoro-4-bromo-chloromethylbenzene, 4-bromo-2-fluorobenzyl chloride, 4-bromo-2-fluorobenzyl chloride, 4-bromo-2-fluorobenzyl chloride
4-bromo-1-(chloromethyl)-2-fluorobenzene

Specifications

1g
C7H5BrClF
UN3265
4-bromo-2-fluorobenzyl chloride, 4-bromo-1-chloromethyl-2-fluorobenzene, 4-bromo-2-fluorobenzylchloride, 4-bromo-1-chloromethyl-2-fluoro-benzene, acmc-209q6a, 4-bromo-2-fluorobenzylchloride, 2-fluoro-4-bromo-chloromethylbenzene, 4-bromo-2-fluorobenzyl chloride, 4-bromo-2-fluorobenzyl chloride, 4-bromo-2-fluorobenzyl chloride
C1=CC(=C(C=C1Br)F)CCl
223.469
223.47
4-Bromo-2-fluorobenzyl chloride
85510-82-3
MFCD02094036
6323063
UDKQGFMDBMYVHI-UHFFFAOYSA-N
4-bromo-1-(chloromethyl)-2-fluorobenzene
2773361
97%
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.