Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-fluorobenzyl alcohol, 97%

$49.73 - $162.73

Chemical Identifiers

Chemical Name or Material 4-Bromo-2-fluorobenzyl alcohol
CAS 188582-62-9
Molecular Formula C7H6BrFO
Formula Weight 205.03
MDL Number MFCD00143265
Beilstein 7757667
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1867906

  SDS
alfa aesar™
L1867906
5g Each for $162.73

AAL1867903

  SDS
alfa aesar™
L1867903
1g Each for $49.73
Description & Specifications

Specifications

Formula Weight 205.03
Health Hazard 1 Warning
MDL Number MFCD00143265
CAS 188582-62-9
Beilstein 7757667
Assay 97%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 4-Bromo-2-fluorobenzyl alcohol
Quantity 5g
Melting Point 31° to 33°C
Molecular Formula C7H6BrFO
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.