Please login to your online account to display your discounted pricing

4-Bromo-2-fluorobenzoyl chloride, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC347460050

 View more versions of this product

Catalog No. AC347460050

  • / Each
 

Description & Specifications

Chemical Identifiers

Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Assay ≥98.5%
Formula Weight 237.46
Physical Form Liquid or Crystalline Mass
Quantity 5g
Color White to light yellow
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 99%
CAS 151982-51-3
CAS Min % 98.5
Packaging Glass bottle
Health Hazard 1 Danger
MDL Number MFCD03411584
Name Note 99%
Molecular Formula C7H3BrClFO
Chemical Name or Material 4-Bromo-2-fluorobenzoyl chloride

Specifications

Chemical Name or Material 4-Bromo-2-fluorobenzoyl chloride
Name Note 99%
CAS 151982-51-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H3BrClFO
Formula Weight 237.46
MDL Number MFCD03411584
Accessories

Danger

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
Causes severe skin burns and eye damage.

SDS & Safety Documents