Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-fluorobenzotrifluoride, 97%

$70.07 - $614.76

Chemical Identifiers

Chemical Name or Material 4-Bromo-2-fluorobenzotrifluoride
CAS 142808-15-9
Molecular Formula C7H3BrF4
Formula Weight 243
MDL Number MFCD01075254
Synonym 3-Fluoro-4-(trifluoromethyl)bromobenzene; 1-Bromo-3-fluoro-4-(trifluoromethyl)benzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2665103

  SDS
alfa aesar™
H2665103
1g Each for $70.07

AAH2665106

  SDS
alfa aesar™
H2665106
5g Each for $203.42

AAH2665114

  SDS
alfa aesar™
H2665114
25g Each for $614.76
Description & Specifications

Specifications

Formula Weight 243
CAS 142808-15-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 4-Bromo-2-fluorobenzotrifluoride
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
MDL Number MFCD01075254
Quantity 1g
Molecular Formula C7H3BrF4
Synonym 3-Fluoro-4-(trifluoromethyl)bromobenzene; 1-Bromo-3-fluoro-4-(trifluoromethyl)benzene
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.