Learn More
4-Bromo-2-fluorobenzonitrile, 99%
CAS: 105942-08-3 | C7H3BrFN | 200.01 g/mol
Supplier: Thermo Scientific Chemicals A1308614
Description
4-Bromo-2-fluorobenzonitrile is used as a OLED intermediate, Pharmaceutical, electronic and chemical intermediate. It is used in the synthesis of heterocycles and liquid crystals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Bromo-2-fluorobenzonitrile is used as a OLED intermediate, Pharmaceutical, electronic and chemical intermediate. It is used in the synthesis of heterocycles and liquid crystals.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications
| 4-Bromo-2-fluorobenzonitrile | |
| 69°C to 72°C | |
| C7H3BrFN | |
| 25 g | |
| 4231143 | |
| Insoluble in water. | |
| FC1=CC(Br)=CC=C1C#N | |
| 200.01 | |
| 200.01 |
| 105942-08-3 | |
| 238°C to 239°C | |
| MFCD00143264 | |
| UN3439 | |
| 2-fluoro-4-bromobenzonitrile, 4-bromo-2-fluoro-benzonitrile, benzonitrile, 4-bromo-2-fluoro, 4-bromo-2-fluorobenzenecarbonitrile, 4-cyano-3-fluorobromobenzene, pubchem3221, 2-floro-4-bromobenzonitrile, 4-bromo-2-fluorobenzontrile, 4-bromo-6-fluorobenzonitrile, ksc490q4t | |
| HGXWRDPQFZKOLZ-UHFFFAOYSA-N | |
| 4-bromo-2-fluorobenzonitrile | |
| 736029 | |
| 99% |
Chemical Identifiers
| 105942-08-3 | |
| 200.01 | |
| HGXWRDPQFZKOLZ-UHFFFAOYSA-N | |
| 736029 | |
| FC1=CC(Br)=CC=C1C#N |
| C7H3BrFN | |
| MFCD00143264 | |
| 2-fluoro-4-bromobenzonitrile, 4-bromo-2-fluoro-benzonitrile, benzonitrile, 4-bromo-2-fluoro, 4-bromo-2-fluorobenzenecarbonitrile, 4-cyano-3-fluorobromobenzene, pubchem3221, 2-floro-4-bromobenzonitrile, 4-bromo-2-fluorobenzontrile, 4-bromo-6-fluorobenzonitrile, ksc490q4t | |
| 4-bromo-2-fluorobenzonitrile |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only