missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-fluorobenzeneboronic acid, 98+%

$141.42 - $590.64

Chemical Identifiers

CAS 216393-64-5
Molecular Formula C6H5BBrFO2
Molecular Weight (g/mol) 218.816
MDL Number MFCD01318110
InChI Key PKZHIIVZRPUZSU-UHFFFAOYSA-N
Synonym 4-bromo-2-fluorobenzeneboronic acid, 4-bromo-2-fluorophenyl boronic acid, 1-borono-4-bromo-2-fluorobenzene, pubchem7778, acmc-209fms, 4-bromo-2-fluorobenzeneboronicacid, 4-bromo-2-fluoro-phenylboronic acid, 4-bromo-2-fluorophenyl boronic acid, 4-bromo-2-fluoro-phenyl boronic acid, boronic acid, 4-bromo-2-fluorophenyl
PubChem CID 2773333
IUPAC Name (4-bromo-2-fluorophenyl)boronic acid
SMILES B(C1=C(C=C(C=C1)Br)F)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1746803 View Documents Alfa Aesar™
L1746803
1g Each for $141.42
 
AAL1746806 View Documents Alfa Aesar™
L1746806
5g Each for $590.64
 
Description

Description

Applications
It is a pharmaceutical intermediate. Arylboronic acids undergo cross-coupling with arenediazonium tetrafluoroborates in dioxane or methanol, catalyzed by palladium acetate in the absence of both added base and phosphine ligand.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

216393-64-5
218.816
PKZHIIVZRPUZSU-UHFFFAOYSA-N
2773333
B(C1=C(C=C(C=C1)Br)F)(O)O
C6H5BBrFO2
MFCD01318110
4-bromo-2-fluorobenzeneboronic acid, 4-bromo-2-fluorophenyl boronic acid, 1-borono-4-bromo-2-fluorobenzene, pubchem7778, acmc-209fms, 4-bromo-2-fluorobenzeneboronicacid, 4-bromo-2-fluoro-phenylboronic acid, 4-bromo-2-fluorophenyl boronic acid, 4-bromo-2-fluoro-phenyl boronic acid, boronic acid, 4-bromo-2-fluorophenyl
(4-bromo-2-fluorophenyl)boronic acid

Specifications

238°C to 242°C
216393-64-5
MFCD01318110
PKZHIIVZRPUZSU-UHFFFAOYSA-N
(4-bromo-2-fluorophenyl)boronic acid
2773333
≥98%
1g
C6H5BBrFO2
4-bromo-2-fluorobenzeneboronic acid, 4-bromo-2-fluorophenyl boronic acid, 1-borono-4-bromo-2-fluorobenzene, pubchem7778, acmc-209fms, 4-bromo-2-fluorobenzeneboronicacid, 4-bromo-2-fluoro-phenylboronic acid, 4-bromo-2-fluorophenyl boronic acid, 4-bromo-2-fluoro-phenyl boronic acid, boronic acid, 4-bromo-2-fluorophenyl
B(C1=C(C=C(C=C1)Br)F)(O)O
218.816
218.82
4-Bromo-2-fluorobenzeneboronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning