Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-fluoroanisole, 97%

$43.86 - $614.18

Chemical Identifiers

Chemical Name or Material 4-Bromo-2-fluoroanisole
CAS 2357-52-0
Molecular Formula C7H6BrFO
Formula Weight 205.03
MDL Number MFCD00011710
Beilstein 3240714
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1094806

 
alfa aesar™
A1094806
5g Each for $43.86

AAA1094814

 
alfa aesar™
A1094814
25g Each for $172.41

AAA1094822

 
alfa aesar™
A1094822
100g Each for $614.18
Description & Specifications

Specifications

Formula Weight 205.03
Chemical Name or Material 4-Bromo-2-fluoroanisole
Assay 97%
Boiling Point 208°C
Quantity 5g
Flash Point 98°C (208°F)
Molecular Formula C7H6BrFO
MDL Number MFCD00011710
CAS 2357-52-0
Beilstein 3240714
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.