missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Bromo-2-fluoroaniline, 99%, ACROS Organics™
Chemical Identifiers
CAS | 367-24-8 |
---|---|
Molecular Formula | C6H5BrFN |
Molecular Weight (g/mol) | 190.01 |
MDL Number | MFCD00010221 |
InChI Key | GZRMNMGWNKSANY-UHFFFAOYSA-N |
Synonym | 2-fluoro-4-bromoaniline, 4-brom-2-fluoranilin, 4-bromo-2-fluoro aniline, 4-bromo-2-fluoro-aniline, 4-bromo-2-fluoro-phenylamine, 4-bromo-2-fluorobenzenamine, 4-bromo-2-fluorophenylamine, benzenamine, 4-bromo-2-fluoro, pubchem1525, zlchem 545 |
PubChem CID | 123050 |
IUPAC Name | 4-bromo-2-fluoroaniline |
SMILES | C1=CC(=C(C=C1Br)F)N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC134180100
|
Thermo Fisher Scientific
AC134180100 |
10g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC134180250
|
Thermo Fisher Scientific
134180250 |
25g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC134181000
|
Thermo Fisher Scientific
134181000 |
100g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
367-24-8 | |
190.01 | |
GZRMNMGWNKSANY-UHFFFAOYSA-N | |
123050 | |
C1=CC(=C(C=C1Br)F)N |
C6H5BrFN | |
MFCD00010221 | |
2-fluoro-4-bromoaniline, 4-brom-2-fluoranilin, 4-bromo-2-fluoro aniline, 4-bromo-2-fluoro-aniline, 4-bromo-2-fluoro-phenylamine, 4-bromo-2-fluorobenzenamine, 4-bromo-2-fluorophenylamine, benzenamine, 4-bromo-2-fluoro, pubchem1525, zlchem 545 | |
4-bromo-2-fluoroaniline |
Specifications
104°C | |
Glass bottle | |
Beige to Brown-Gray or Gray | |
10g | |
98.5 | |
98.5% min. (GC) | |
BrC6H3(F)NH2 | |
2-fluoro-4-bromoaniline, 4-brom-2-fluoranilin, 4-bromo-2-fluoro aniline, 4-bromo-2-fluoro-aniline, 4-bromo-2-fluoro-phenylamine, 4-bromo-2-fluorobenzenamine, 4-bromo-2-fluorophenylamine, benzenamine, 4-bromo-2-fluoro, pubchem1525, zlchem 545 | |
C1=CC(=C(C=C1Br)F)N | |
190.01 | |
190.01 | |
99% |
Authentic | |
103°C to 108°C (13.0mmHg) | |
39°C to 42°C | |
367-24-8 | |
100.0 | |
C6H5BrFN | |
MFCD00010221 | |
GZRMNMGWNKSANY-UHFFFAOYSA-N | |
4-bromo-2-fluoroaniline | |
123050 | |
Crystalline Mass, Powder, Crystals and/or Chunks | |
4-Bromo-2-fluoroaniline, 99% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
GHS Signal Word: Warning
EINECSNumber : 206-685-4