missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-fluoroaniline, 98+%

Manufacturer:  Alfa Aesar™ A1022822

 View more versions of this product

Catalog No. AAA1022822



Specifications

Specifications

4-Bromo-2-fluoroaniline
103°C to 108°C (13mmHg)
100g
C6H5BrFN
UN2811
2-fluoro-4-bromoaniline, benzenamine, 4-bromo-2-fluoro, 4-bromo-2-fluoro-phenylamine, 4-bromo-2-fluoro-aniline, 4-bromo-2-fluorobenzenamine, 4-bromo-2-fluoro aniline, 4-bromo-2-fluorophenylamine, 4-brom-2-fluoranilin, zlchem 545, pubchem1525
C1=CC(=C(C=C1Br)F)N
190.015
190.01
104°C (219°F)
38°C to 42°C
367-24-8
MFCD00010221
2081089
GZRMNMGWNKSANY-UHFFFAOYSA-N
4-bromo-2-fluoroaniline
123050
≥98%

Chemical Identifiers

367-24-8
190.015
GZRMNMGWNKSANY-UHFFFAOYSA-N
123050
C1=CC(=C(C=C1Br)F)N
C6H5BrFN
MFCD00010221
2-fluoro-4-bromoaniline, benzenamine, 4-bromo-2-fluoro, 4-bromo-2-fluoro-phenylamine, 4-bromo-2-fluoro-aniline, 4-bromo-2-fluorobenzenamine, 4-bromo-2-fluoro aniline, 4-bromo-2-fluorophenylamine, 4-brom-2-fluoranilin, zlchem 545, pubchem1525
4-bromo-2-fluoroaniline
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

SDS Documents