Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Bromo-2'-fluoroacetophenone, 96%

Chemical Identifiers

Chemical Name or Material 4'-Bromo-2'-fluoroacetophenone
CAS 625446-22-2
Formula Weight 217.04
MDL Number MFCD03411551
Synonym 1-(4-Bromo-2-fluorophenyl)ethanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6212703

 
alfa aesar™
H6212703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6212706

 
alfa aesar™
H6212706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62127MD

 
alfa aesar™
H62127MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 217.04
Chemical Name or Material 4'-Bromo-2'-fluoroacetophenone
Physical Form Solid
Quantity 1g
MDL Number MFCD03411551
Assay 96%
Synonym 1-(4-Bromo-2-fluorophenyl)ethanone
CAS 625446-22-2