Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Bromo-2'-fluoroacetanilide, 98%

$37.68 - $572.57

Chemical Identifiers

Chemical Name or Material 4'-Bromo-2'-fluoroacetanilide
CAS 326-66-9
Molecular Formula C8H7BrFNO
Formula Weight 232.05
MDL Number MFCD00042588
Beilstein 3250459
Synonym N-(4-Bromo-2-fluorophenyl)acetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1572309

  SDS
alfa aesar™
A1572309
10g Each for $37.68

AAA1572318

  SDS
alfa aesar™
A1572318
50g Each for $153.69

AAA1572330

  SDS
alfa aesar™
A1572330
250g Each for $572.57
Description & Specifications

Specifications

Formula Weight 232.05
Chemical Name or Material 4'-Bromo-2'-fluoroacetanilide
Assay 98%
Molecular Formula C8H7BrFNO
MDL Number MFCD00042588
CAS 326-66-9
Melting Point 154° to 158°C
Beilstein 3250459
Synonym N-(4-Bromo-2-fluorophenyl)acetamide
Quantity 10g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.