missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Bromo-2'-fluoroacetanilide, 98%

Manufacturer:  Alfa Aesar™ A1572318

 View more versions of this product

Catalog No. AAA1572318



Specifications

Specifications

4'-Bromo-2'-fluoroacetanilide
50g
C8H7BrFNO
3250459
BCYGKMDWQBWUSC-UHFFFAOYSA-N
N-(4-bromo-2-fluorophenyl)acetamide
67593
98%
154°C to 158°C
326-66-9
MFCD00042588
n-4-bromo-2-fluorophenyl acetamide, 4'-bromo-2'-fluoroacetanilide, 4-bromo-2-fluoroacetanilide, acetamide, n-4-bromo-2-fluorophenyl, 4-bromo-2-fluoroacetanilide, pubchem14886, acmc-1cooc, ksc497m2b, timtec-bb sbb009911, attercop-chm at130693
CC(=O)NC1=C(C=C(C=C1)Br)F
232.052
232.05

Chemical Identifiers

326-66-9
232.052
BCYGKMDWQBWUSC-UHFFFAOYSA-N
67593
CC(=O)NC1=C(C=C(C=C1)Br)F
C8H7BrFNO
MFCD00042588
n-4-bromo-2-fluorophenyl acetamide, 4'-bromo-2'-fluoroacetanilide, 4-bromo-2-fluoroacetanilide, acetamide, n-4-bromo-2-fluorophenyl, 4-bromo-2-fluoroacetanilide, pubchem14886, acmc-1cooc, ksc497m2b, timtec-bb sbb009911, attercop-chm at130693
N-(4-bromo-2-fluorophenyl)acetamide
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents