Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-fluoro-1-(trifluoromethoxy)benzene, 97%

$119.45 - $661.89

Chemical Identifiers

Chemical Name or Material 4-Bromo-2-fluoro-1-(trifluoromethoxy)benzene
CAS 105529-58-6
Molecular Formula C7H3BrF4O
Formula Weight 259
MDL Number MFCD01861132
Synonym 1-Bromo-3-fluoro-4-(trifluoromethoxy)benzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2649903

 
alfa aesar™
H2649903
1g Each for $119.45

AAH2649909

 
alfa aesar™
H2649909
10g Each for $661.89
Description & Specifications

Specifications

Formula Weight 259
Chemical Name or Material 4-Bromo-2-fluoro-1-(trifluoromethoxy)benzene
Assay 97%
Molecular Formula C7H3BrF4O
MDL Number MFCD01861132
CAS 105529-58-6
Synonym 1-Bromo-3-fluoro-4-(trifluoromethoxy)benzene
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.