missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-2-fluoro-1-nitrobenzene, TCI America™

$38.19 - $110.00

Chemical Identifiers

CAS 321-23-3
Molecular Formula C6H3BrFNO2
Molecular Weight (g/mol) 219.997
MDL Number MFCD01930221
InChI Key VQCWSOYHHXXWSP-UHFFFAOYSA-N
Synonym 4-bromo-2-fluoronitrobenzene, 2-fluoro-4-bromonitrobenzene, 4-bromo-2-fluoro-1-nitro-benzene, benzene, 4-bromo-2-fluoro-1-nitro, 1-bromo-3-fluoro-4-nitrobenzene, 2-fluoro-4-bromo-nitrobenzene, 4-bromo-2-fluoro-nitrobenzene, 4-bromo-2-fluornitrobenzole, 3-fluoro-4-nitrobromobenzene, 5-bromo-2-nitrofluorobenzene
PubChem CID 2783362
IUPAC Name 4-bromo-2-fluoro-1-nitrobenzene
SMILES C1=CC(=C(C=C1Br)F)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B30625G View Documents TCI America
B30625G
5g Each for $38.19
 
B306225G View Documents TCI America
B306225G
25g Each for $110.00
 
Specifications

Chemical Identifiers

321-23-3
219.997
VQCWSOYHHXXWSP-UHFFFAOYSA-N
2783362
C1=CC(=C(C=C1Br)F)[N+](=O)[O-]
C6H3BrFNO2
MFCD01930221
4-bromo-2-fluoronitrobenzene, 2-fluoro-4-bromonitrobenzene, 4-bromo-2-fluoro-1-nitro-benzene, benzene, 4-bromo-2-fluoro-1-nitro, 1-bromo-3-fluoro-4-nitrobenzene, 2-fluoro-4-bromo-nitrobenzene, 4-bromo-2-fluoro-nitrobenzene, 4-bromo-2-fluornitrobenzole, 3-fluoro-4-nitrobromobenzene, 5-bromo-2-nitrofluorobenzene
4-bromo-2-fluoro-1-nitrobenzene

Specifications

Undesignated
5g
C6H3BrFNO2
4-bromo-2-fluoronitrobenzene, 2-fluoro-4-bromonitrobenzene, 4-bromo-2-fluoro-1-nitro-benzene, benzene, 4-bromo-2-fluoro-1-nitro, 1-bromo-3-fluoro-4-nitrobenzene, 2-fluoro-4-bromo-nitrobenzene, 4-bromo-2-fluoro-nitrobenzene, 4-bromo-2-fluornitrobenzole, 3-fluoro-4-nitrobromobenzene, 5-bromo-2-nitrofluorobenzene
C1=CC(=C(C=C1Br)F)[N+](=O)[O-]
219.997
220.00
≥98.0% (GC)
86°C
321-23-3
MFCD01930221
VQCWSOYHHXXWSP-UHFFFAOYSA-N
4-bromo-2-fluoro-1-nitrobenzene
2783362
Crystal-Powder at 20°C
4-Bromo-2-fluoro-1-nitrobenzene
Safety and Handling

Safety and Handling

TSCA : No