Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-ethylaniline hydrochloride, 98%

$60.80 - $207.80

Chemical Identifiers

Chemical Name or Material 4-Bromo-2-ethylaniline hydrochloride
CAS 30273-22-4
Molecular Formula C8H10BrN·HCl
Formula Weight 236.55
MDL Number MFCD00156127
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1307706

 
alfa aesar™
A1307706
5g Each for $60.80

AAA1307714

 
alfa aesar™
A1307714
25g Each for $207.80
Description & Specifications

Specifications

Formula Weight 236.55
Chemical Name or Material 4-Bromo-2-ethylaniline hydrochloride
Assay 98%
Molecular Formula C8H10BrN·HCl
MDL Number MFCD00156127
CAS 30273-22-4
Melting Point ≈222°C (dec.)
Quantity 5g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.