missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-2-chlorotoluene, 98%, ACROS Organics™

Chemical Identifiers

CAS 89794-02-5
Molecular Formula C7H6BrCl
Molecular Weight (g/mol) 205.48
MDL Number MFCD00060651
InChI Key LIFMTDJMLRECMX-UHFFFAOYSA-N
Synonym 2-chloro 4-bromo toluene, 2-chloro-4-bromotoluene, 4-bromo-2-chloro toluene, 4-bromo-2-chloro-1-methyl-benzene, 4-bromo-2-chlorotoluene, 4-bromo-2chlorotoluene, 4-bromo-chlorotoluene, acmc-209r2x, benzene, 4-bromo-2-chloro-1-methyl, pubchem3603
PubChem CID 2735554
IUPAC Name 4-bromo-2-chloro-1-methylbenzene
SMILES CC1=C(C=C(C=C1)Br)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC398280250
SDS View Documents
Thermo Fisher Scientific
AC398280250
25g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC398281000
SDS View Documents
Thermo Fisher Scientific
AC398281000
100g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

89794-02-5
205.48
LIFMTDJMLRECMX-UHFFFAOYSA-N
2735554
CC1=C(C=C(C=C1)Br)Cl
C7H6BrCl
MFCD00060651
2-chloro 4-bromo toluene, 2-chloro-4-bromotoluene, 4-bromo-2-chloro toluene, 4-bromo-2-chloro-1-methyl-benzene, 4-bromo-2-chlorotoluene, 4-bromo-2chlorotoluene, 4-bromo-chlorotoluene, acmc-209r2x, benzene, 4-bromo-2-chloro-1-methyl, pubchem3603
4-bromo-2-chloro-1-methylbenzene

Specifications

1.5380g/mL
Authentic
1.5730 to 1.5750 (20°C, 589nm)
212°C
25g
97.5
97.5% min. (GC)
BrC6H3(Cl)CH3
2-chloro 4-bromo toluene, 2-chloro-4-bromotoluene, 4-bromo-2-chloro toluene, 4-bromo-2-chloro-1-methyl-benzene, 4-bromo-2-chlorotoluene, 4-bromo-2chlorotoluene, 4-bromo-chlorotoluene, acmc-209r2x, benzene, 4-bromo-2-chloro-1-methyl, pubchem3603
CC1=C(C=C(C=C1)Br)Cl
205.48
205.48
98%
95°C
Glass bottle
1.538
Yellow
89794-02-5
100.0
C7H6BrCl
MFCD00060651
LIFMTDJMLRECMX-UHFFFAOYSA-N
4-bromo-2-chloro-1-methylbenzene
2735554
Liquid
4-Bromo-2-chlorotoluene
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning