missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-2-chloropyridine, TCI America™

$39.72 - $117.64

Chemical Identifiers

CAS 73583-37-6
Molecular Formula C5H3BrClN
Molecular Weight (g/mol) 192.44
MDL Number MFCD03840756
InChI Key ONHMWUXYIFULDO-UHFFFAOYSA-N
Synonym 2-chloro-4-bromopyridine, pyridine, 4-bromo-2-chloro, 4-bromo-2-chloro-pyridine, pubchem1171, acmc-1bjmw, ksc381c4t, tpc-py061, 4-bromo-2-chloropyridine, abbypharma ap-18-5387, 4-bromo-2-chloropyridine 5g
PubChem CID 3847770
IUPAC Name 4-bromo-2-chloropyridine
SMILES C1=CN=C(C=C1Br)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B29511G View Documents TCI America
B29511G
1g Each for $39.72
 
B29515G View Documents TCI America
B29515G
5g Each for $117.64
 
Specifications

Chemical Identifiers

73583-37-6
192.44
ONHMWUXYIFULDO-UHFFFAOYSA-N
3847770
C1=CN=C(C=C1Br)Cl
C5H3BrClN
MFCD03840756
2-chloro-4-bromopyridine, pyridine, 4-bromo-2-chloro, 4-bromo-2-chloro-pyridine, pubchem1171, acmc-1bjmw, ksc381c4t, tpc-py061, 4-bromo-2-chloropyridine, abbypharma ap-18-5387, 4-bromo-2-chloropyridine 5g
4-bromo-2-chloropyridine

Specifications

70°C
1g
C5H3BrClN
2-chloro-4-bromopyridine, pyridine, 4-bromo-2-chloro, 4-bromo-2-chloro-pyridine, pubchem1171, acmc-1bjmw, ksc381c4t, tpc-py061, 4-bromo-2-chloropyridine, abbypharma ap-18-5387, 4-bromo-2-chloropyridine 5g
C1=CN=C(C=C1Br)Cl
192.44
192.44
≥98.0% (GC)
Yellow
73583-37-6
MFCD03840756
ONHMWUXYIFULDO-UHFFFAOYSA-N
4-bromo-2-chloropyridine
3847770
Clear Liquid at 20°C
4-Bromo-2-chloropyridine
Safety and Handling

Safety and Handling

TSCA : No