missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-2-chloropyridine, 97%, ACROS Organics™

Chemical Identifiers

CAS 73583-37-6
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C5H3BrClN
Formula Weight 192.44
MDL Number MFCD03840756
Chemical Name or Material 4-Bromo-2-chloropyridine
Assay ≥96.0% (GC)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC434800050 SDS Thermo Fisher Scientific
AC434800050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC434800250 SDS Thermo Fisher Scientific
AC434800250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

73583-37-6
100.0
C5H3BrClN
MFCD03840756
≥96.0% (GC)
96.0
97%
192.44
4-Bromo-2-chloropyridine

Specifications

Powder
5g
1.734g/mL
96.0
C5H3BrClN
73583-37-6
Authentic
≥96.0% (GC)
192.44
Yellow
100.0
Glass bottle
97%
MFCD03840756
4-Bromo-2-chloropyridine

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.