missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-2-chlorobenzotrifluoride, TCI America™

$85.56 - $253.61

Chemical Identifiers

CAS 467435-07-0
Molecular Formula C7H3BrClF3
Molecular Weight (g/mol) 259.45
MDL Number MFCD08459292
InChI Key FJANWAMBKVZTRG-UHFFFAOYSA-N
Synonym 4-Bromo-2-chloro-1-(trifluoromethyl)benzene, 4-Bromo-2-chloro-alpha,alpha,alpha-trifluorotoluene
PubChem CID 20269848
IUPAC Name 4-bromo-2-chloro-1-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1Br)Cl)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B30575G View Documents TCI America
B30575G
5g Each for $85.56
 
B305725G View Documents TCI America
B305725G
25g Each for $253.61
 
Specifications

Chemical Identifiers

467435-07-0
259.45
FJANWAMBKVZTRG-UHFFFAOYSA-N
20269848
C1=CC(=C(C=C1Br)Cl)C(F)(F)F
C7H3BrClF3
MFCD08459292
4-Bromo-2-chloro-1-(trifluoromethyl)benzene, 4-Bromo-2-chloro-alpha,alpha,alpha-trifluorotoluene
4-bromo-2-chloro-1-(trifluoromethyl)benzene

Specifications

Yellow
467435-07-0
MFCD08459292
FJANWAMBKVZTRG-UHFFFAOYSA-N
4-bromo-2-chloro-1-(trifluoromethyl)benzene
20269848
Clear Liquid at 20°C
4-Bromo-2-chlorobenzotrifluoride
5g
C7H3BrClF3
4-Bromo-2-chloro-1-(trifluoromethyl)benzene, 4-Bromo-2-chloro-alpha,alpha,alpha-trifluorotoluene
C1=CC(=C(C=C1Br)Cl)C(F)(F)F
259.45
259.45
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No