missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-chlorobenzonitrile, 96%

Manufacturer:  Alfa Aesar™ L1060206

 View more versions of this product

Catalog No. AAL1060206



Specifications

Specifications

4-Bromo-2-chlorobenzonitrile
67°C to 72°C
154607-01-9
MFCD00040883
3240885
AYQBMZNSJPVADT-UHFFFAOYSA-N
4-bromo-2-chlorobenzonitrile
7010420
96%
142°C to 143°C (10mmHg)
5g
C7H3BrClN
UN3439
2-chloro-4-bromobenzonitrile, 4-bromo-2-chloro-benzonitrile, benzonitrile, 4-bromo-2-chloro, 4-bromo-2-chlorobenzenecarbonitrile, pubchem12801, intermediates-zcf02177, acmc-209db5, ksc497g8d
C1=CC(=C(C=C1Br)Cl)C#N
216.462
216.47

Chemical Identifiers

154607-01-9
216.462
AYQBMZNSJPVADT-UHFFFAOYSA-N
7010420
C1=CC(=C(C=C1Br)Cl)C#N
C7H3BrClN
MFCD00040883
2-chloro-4-bromobenzonitrile, 4-bromo-2-chloro-benzonitrile, benzonitrile, 4-bromo-2-chloro, 4-bromo-2-chlorobenzenecarbonitrile, pubchem12801, intermediates-zcf02177, acmc-209db5, ksc497g8d
4-bromo-2-chlorobenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Documents